siRNA Details
| Gene Symbol: Pkig | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153469.4 | NP_703199.1 | 3 | 280 | ||
| XM_006235518.2 | XP_006235580.1 | 3 | 1521 | |||
| XM_006235520.2 | XP_006235582.1 | 4 | 372 | |||
| XM_006235521.1 | XP_006235583.1 | 4 | 464 | |||
| XM_006235522.1 | XP_006235584.1 | 3 | 395 | |||
| XM_008762469.1 | XP_008760691.1 | 3 | 1742 | |||
| XM_008762470.1 | XP_008760692.1 | 3 | 311 | |||
| GenBank mRNA | AY150308.3 | - | 3 | 252 | ||
| BC088108.1 | - | 3 | 280 | 7368431 | ||
| FQ218619.1 | - | 3 | 376 | |||

Pkig
Genomic Map