siRNA Details
Gene Symbol: Pkig | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153469.4 | NP_703199.1 | 2 | 94 | ||
XM_006235518.2 | XP_006235580.1 | 2 | 1335 | |||
XM_006235520.2 | XP_006235582.1 | 3 | 186 | |||
XM_006235521.1 | XP_006235583.1 | 3 | 278 | |||
XM_006235522.1 | XP_006235584.1 | 2 | 209 | |||
XM_008762469.1 | XP_008760691.1 | 2 | 1556 | |||
XM_008762470.1 | XP_008760692.1 | 2 | 125 | |||
GenBank mRNA | AY150308.3 | - | 2 | 66 | ||
BC088108.1 | - | 2 | 94 | 7368431 | ||
FQ218619.1 | - | 2 | 190 |
