siRNA Details
| Gene Symbol: Mnat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153472.2 | NP_703202.1 | 7 | 758 | ||
| GenBank mRNA | AY135567.1 | - | 7 | 713 | ||
| BC078712.1 | - | 7 | 758 | 7128354 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mnat1| Gene Symbol: Mnat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153472.2 | NP_703202.1 | 7 | 758 | ||
| GenBank mRNA | AY135567.1 | - | 7 | 713 | ||
| BC078712.1 | - | 7 | 758 | 7128354 | ||