siRNA Details
| Gene Symbol: Slc12a8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153625.1 | NP_705889.1 | 9 | 1488 | ||
| GenBank mRNA | AB024430.1 | - | 9 | 1488 | ||
| AB024431.1 | - | 9 | 1410 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc12a8| Gene Symbol: Slc12a8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153625.1 | NP_705889.1 | 9 | 1488 | ||
| GenBank mRNA | AB024430.1 | - | 9 | 1488 | ||
| AB024431.1 | - | 9 | 1410 | |||