siRNA Details
| Gene Symbol: Prop1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153627.1 | NP_705891.1 | 1 | 98 | ||
| XM_008767664.1 | XP_008765886.1 | 1 | 95 | |||
| GenBank mRNA | AB037922.1 | - | 1 | 98 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prop1| Gene Symbol: Prop1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153627.1 | NP_705891.1 | 1 | 98 | ||
| XM_008767664.1 | XP_008765886.1 | 1 | 95 | |||
| GenBank mRNA | AB037922.1 | - | 1 | 98 | ||