siRNA Details
| Gene Symbol: Nptx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153735.2 | NP_714957.1 | 3 | 900 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nptx1| Gene Symbol: Nptx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153735.2 | NP_714957.1 | 3 | 900 | ||