siRNA Details
| Gene Symbol: Has3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172319.1 | NP_758822.1 | 4 | 1765 | ||
| XM_006255561.2 | XP_006255623.1 | 4 | 2985 | |||
| GenBank mRNA | AB097569.1 | - | 4 | 1765 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Has3| Gene Symbol: Has3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172319.1 | NP_758822.1 | 4 | 1765 | ||
| XM_006255561.2 | XP_006255623.1 | 4 | 2985 | |||
| GenBank mRNA | AB097569.1 | - | 4 | 1765 | ||