siRNA Details
| Gene Symbol: Duox1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153739.1 | NP_714961.1 | 24, 25 | 3442 | ||
| XM_006234829.2 | XP_006234891.1 | 25, 26 | 3541 | |||
| GenBank mRNA | AF542180.1 | - | 24, 25 | 3442 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Duox1| Gene Symbol: Duox1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153739.1 | NP_714961.1 | 24, 25 | 3442 | ||
| XM_006234829.2 | XP_006234891.1 | 25, 26 | 3541 | |||
| GenBank mRNA | AF542180.1 | - | 24, 25 | 3442 | ||