siRNA Details
| Gene Symbol: Rln3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_170667.2 | NP_733767.1 | 1 | 139 | ||
| GenBank mRNA | AB076564.1 | - | 1 | 139 | ||
| AY112741.1 | - | 1 | 139 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rln3| Gene Symbol: Rln3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_170667.2 | NP_733767.1 | 1 | 139 | ||
| GenBank mRNA | AB076564.1 | - | 1 | 139 | ||
| AY112741.1 | - | 1 | 139 | |||