siRNA Details
Gene Symbol: Rln3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_170667.2 | NP_733767.1 | 2 | 289 | ||
GenBank mRNA | AB076564.1 | - | 2 | 289 | ||
AY112741.1 | - | 2 | 289 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Rln3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_170667.2 | NP_733767.1 | 2 | 289 | ||
GenBank mRNA | AB076564.1 | - | 2 | 289 | ||
AY112741.1 | - | 2 | 289 |