siRNA Details
| Gene Symbol: Gfer | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013222.2 | NP_037354.2 | 1, 2 | 320 | ||
| GenBank mRNA | D30735.1 | - | 1, 2 | 320 | ||
| Gene Symbol: LOC100912596 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003752199.3 | XP_003752247.2 | 1, 2 | 1534 | ||

Gfer
Genomic Map