siRNA Details
| Gene Symbol: Wnt5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100489.1 | NP_001093959.1 | 4 | 565 | ||
| XM_006237179.1 | XP_006237241.1 | 3 | 443 | |||
| GenBank mRNA | AF481944.1 | - | 2 | 90 | ||
| BC161860.1 | - | 708 | 7096336 | |||

Wnt5b
Genomic Map