siRNA Details
| Gene Symbol: Mrfap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009264.1 | NP_001009264.1 | 1 | 432 | ||
| GenBank mRNA | AF526268.1 | - | 2 | 382 | ||
| BC088308.1 | - | 1 | 432 | 7324285 | ||
| FQ228750.1 | - | 1 | 452 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mrfap1| Gene Symbol: Mrfap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009264.1 | NP_001009264.1 | 1 | 432 | ||
| GenBank mRNA | AF526268.1 | - | 2 | 382 | ||
| BC088308.1 | - | 1 | 432 | 7324285 | ||
| FQ228750.1 | - | 1 | 452 | |||