siRNA Details
Gene Symbol: Ppif | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172243.1 | NP_758443.1 | 1, 2 | 212 | ||
GenBank mRNA | BC086977.1 | - | 1, 2 | 245 | 7193156 | |
U68544.1 | - | 1, 2 | 212 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ppif | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172243.1 | NP_758443.1 | 1, 2 | 212 | ||
GenBank mRNA | BC086977.1 | - | 1, 2 | 245 | 7193156 | |
U68544.1 | - | 1, 2 | 212 |