siRNA Details
Gene Symbol: Wrnip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172332.2 | NP_758835.1 | 7 | 2068 | ||
GenBank mRNA | AY136827.1 | - | 7 | 1972 | ||
BC098652.1 | - | 7 | 2068 | 7306780 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Wrnip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172332.2 | NP_758835.1 | 7 | 2068 | ||
GenBank mRNA | AY136827.1 | - | 7 | 1972 | ||
BC098652.1 | - | 7 | 2068 | 7306780 |