siRNA Details
| Gene Symbol: Dpy30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170545.1 | NP_001164016.1 | 4 | 243 | ||
| NM_173117.2 | NP_775140.2 | 4 | 282 | |||
| GenBank mRNA | AY129401.1 | - | 4 | 282 | ||
| FM121127.1 | - | 4 | 249 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dpy30| Gene Symbol: Dpy30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170545.1 | NP_001164016.1 | 4 | 243 | ||
| NM_173117.2 | NP_775140.2 | 4 | 282 | |||
| GenBank mRNA | AY129401.1 | - | 4 | 282 | ||
| FM121127.1 | - | 4 | 249 | |||