siRNA Details
| Gene Symbol: Vimp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173120.2 | NP_775143.2 | 6 | 587 | ||
| GenBank mRNA | AF367467.1 | - | 6 | 597 | ||
| BC059111.1 | - | 6 | 591 | 6920258 | ||
| FQ210095.1 | - | 6 | 579 | |||
| FQ218853.1 | - | 6 | 597 | |||
| FQ219944.1 | - | 6 | 579 | |||
| FQ220082.1 | - | 6 | 582 | |||
| FQ225575.1 | - | 6 | 598 | |||
| FQ229642.1 | - | 6 | 579 | |||

Vimp
Genomic Map