siRNA Details
| Gene Symbol: Smim3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173126.3 | NP_775149.1 | 2 | 412 | ||
| XM_006254762.2 | XP_006254824.1 | 4 | 1095 | |||
| XM_006254763.2 | XP_006254825.1 | 4 | 1098 | |||
| XM_006254765.2 | XP_006254827.1 | 4 | 624 | |||
| XM_006254767.1 | XP_006254829.1 | 2 | 98 | |||
| XM_006254769.2 | XP_006254831.1 | 2 | 301 | |||
| XM_008772096.1 | XP_008770318.1 | 3 | 1226 | |||
| XM_008772097.1 | XP_008770319.1 | 2 | 1034 | |||
| GenBank mRNA | BC070515.1 | - | 2 | 412 | 7100997 | |
| EV780394.1 | - | 2 | 193 | |||

Smim3
Genomic Map