siRNA Details
| Gene Symbol: Prss3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173127.1 | NP_775150.1 | 2 | 99 | ||
| GenBank mRNA | FQ226997.1 | - | 2 | 130 | ||
| FQ230379.1 | - | 2 | 130 | |||
| FQ235277.1 | - | 2 | 130 | |||
| M16624.1 | - | 2 | 99 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prss3| Gene Symbol: Prss3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173127.1 | NP_775150.1 | 2 | 99 | ||
| GenBank mRNA | FQ226997.1 | - | 2 | 130 | ||
| FQ230379.1 | - | 2 | 130 | |||
| FQ235277.1 | - | 2 | 130 | |||
| M16624.1 | - | 2 | 99 | |||