siRNA Details
| Gene Symbol: Vom2r27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173130.1 | NP_775153.1 | 2 | 390 | ||
| XM_006228015.1 | XP_006228077.1 | 2 | 392 | |||
| GenBank mRNA | AF016182.1 | - | 2 | 390 | ||
More Information
Set Membership: |
|

Vom2r27
Genomic Map