siRNA Details
| Gene Symbol: Vom2r53 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173131.2 | NP_775154.1 | 6 | 2551 | ||
| GenBank mRNA | AF016178.1 | - | 6 | 2551 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vom2r53| Gene Symbol: Vom2r53 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173131.2 | NP_775154.1 | 6 | 2551 | ||
| GenBank mRNA | AF016178.1 | - | 6 | 2551 | ||
Set Membership: |
|