siRNA Details
Gene Symbol: Cryba2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173140.1 | NP_775163.1 | 5 | 520 | ||
GenBank mRNA | AY158891.1 | - | 5 | 520 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Cryba2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173140.1 | NP_775163.1 | 5 | 520 | ||
GenBank mRNA | AY158891.1 | - | 5 | 520 |