siRNA Details
| Gene Symbol: Stk16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173142.1 | NP_775165.1 | 6 | 876 | ||
| GenBank mRNA | D86220.1 | - | 6 | 876 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Stk16| Gene Symbol: Stk16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173142.1 | NP_775165.1 | 6 | 876 | ||
| GenBank mRNA | D86220.1 | - | 6 | 876 | ||