siRNA Details
| Gene Symbol: Gimap4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173153.2 | NP_775176.2 | 2, 3 | 104 | ||
| GenBank mRNA | AM285343.1 | - | 2, 3 | 104 | ||
| AM285683.1 | - | 2, 3 | 104 | |||
| AY070268.1 | - | 1, 2 | 86 | |||
| BC070952.1 | - | 2, 3 | 92 | 7100695 | ||
| CK479589.1 | - | 2, 3 | 104 | |||
| DQ125339.1 | - | 2, 3 | 93 | |||
| DQ125340.1 | - | 2, 3 | 93 | |||

Gimap4
Genomic Map