siRNA Details
| Gene Symbol: Chst15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173310.3 | NP_775432.3 | 1 | 267 | ||
| XM_006230430.2 | XP_006230492.1 | 2 | 1834 | |||
| XM_006230431.2 | XP_006230493.1 | 2 | 2574 | |||
| XM_006230433.2 | XP_006230495.1 | 2 | 2069 | |||
| XM_008759868.1 | XP_008758090.1 | 2 | 830 | |||
| XM_008759869.1 | XP_008758091.1 | 2 | 2573 | |||
| GenBank mRNA | AB086953.1 | - | 1 | 267 | ||
| BC085687.1 | - | 1 | 528 | 7189319 | ||

Chst15
Genomic Map