siRNA Details
| Gene Symbol: Mapk15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173331.2 | NP_775453.1 | 14 | 1554 | ||
| XM_006241777.2 | XP_006241839.1 | 14 | 2078 | |||
| XM_006241779.2 | XP_006241841.1 | 15 | 1970 | |||
| XM_008765547.1 | XP_008763769.1 | 14 | 1930 | |||
| GenBank mRNA | AF078798.2 | - | 14 | 1670 | ||
| FQ212754.1 | - | 9 | 1434 | |||

Mapk15
Genomic Map