siRNA Details
| Gene Symbol: Slco6c1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173338.1 | NP_775460.1 | 346 | |||
| GenBank mRNA | AF326113.1 | - | 346 | |||
| Gene Symbol: LOC102554041 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008767397.1 | XP_008765619.1 | 1, 2 | 515 | ||

Slco6c1
Genomic Map