siRNA Details
| Gene Symbol: Mettl22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_213210.8 | XP_213210.4 | 3 | 397 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mettl22| Gene Symbol: Mettl22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_213210.8 | XP_213210.4 | 3 | 397 | ||