siRNA Details
| Gene Symbol: Olr1365 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_214824.1 | NP_999989.1 | 493 | |||
| Gene Symbol: LOC100912159 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003750781.2 | XP_003750829.2 | 2 | 534 | ||

Olr1365
Genomic Map