siRNA Details
| Gene Symbol: Zfp213 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105764.1 | NP_001099234.1 | 2 | 536 | ||
| XM_006245862.2 | XP_006245924.1 | 2 | 553 | |||
| XM_006245863.2 | XP_006245925.1 | 2 | 1089 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp213| Gene Symbol: Zfp213 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105764.1 | NP_001099234.1 | 2 | 536 | ||
| XM_006245862.2 | XP_006245924.1 | 2 | 553 | |||
| XM_006245863.2 | XP_006245925.1 | 2 | 1089 | |||