siRNA Details
Gene Symbol: Zfp213 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105764.1 | NP_001099234.1 | 3 | 818 | ||
XM_006245862.2 | XP_006245924.1 | 3 | 835 | |||
XM_006245863.2 | XP_006245925.1 | 3 | 1371 |
