siRNA Details
| Gene Symbol: Cldn9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011889.1 | NP_001011889.1 | 1 | 381 | ||
| GenBank mRNA | BC087664.1 | - | 1 | 381 | 7315462 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cldn9| Gene Symbol: Cldn9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011889.1 | NP_001011889.1 | 1 | 381 | ||
| GenBank mRNA | BC087664.1 | - | 1 | 381 | 7315462 | |