siRNA Details
| Gene Symbol: Kremen2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105767.1 | NP_001099237.1 | 7 | 858 | ||
| XM_008767550.1 | XP_008765772.1 | 6 | 1081 | |||
| XM_008767551.1 | XP_008765773.1 | 6 | 1081 | |||
| XM_008767552.1 | XP_008765774.1 | 6 | 1081 | |||

Kremen2
Genomic Map