siRNA Details
| Gene Symbol: Fam173a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127447.1 | NP_001120919.1 | 1 | 185 | ||
| GenBank mRNA | AW918226.1 | - | 1 | 185 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam173a| Gene Symbol: Fam173a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127447.1 | NP_001120919.1 | 1 | 185 | ||
| GenBank mRNA | AW918226.1 | - | 1 | 185 | ||