siRNA Details
| Gene Symbol: Mrpl55 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105782.1 | NP_001099252.1 | 3 | 401 | ||
| XM_006246446.2 | XP_006246508.1 | 3 | 442 | |||
| XM_006246447.2 | XP_006246509.1 | 3 | 432 | |||
| XM_008767780.1 | XP_008766002.1 | 3 | 794 | |||

Mrpl55
Genomic Map