siRNA Details
| Gene Symbol: Wnt9a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105783.1 | NP_001099253.1 | 3 | 670 | ||
| XM_006246448.2 | XP_006246510.1 | 3 | 680 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Wnt9a| Gene Symbol: Wnt9a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105783.1 | NP_001099253.1 | 3 | 670 | ||
| XM_006246448.2 | XP_006246510.1 | 3 | 680 | |||