siRNA Details
| Gene Symbol: Zfp496 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271396.1 | NP_001258325.1 | 10 | 1899 | ||
| XM_006246450.1 | XP_006246512.1 | 10 | 2048 | |||
| XM_006246451.1 | XP_006246513.1 | 10 | 1890 | |||
| XM_006246452.1 | XP_006246514.1 | 10 | 1904 | |||
| XM_008767781.1 | XP_008766003.1 | 7 | 1200 | |||
| GenBank mRNA | FQ223523.1 | - | 10 | 1901 | ||

Zfp496
Genomic Map