siRNA Details
| Gene Symbol: B9d1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105786.1 | NP_001099256.1 | 1, 2 | 111 | ||
| XM_006246553.2 | XP_006246615.1 | 817 | ||||
| XM_006246554.2 | XP_006246616.1 | 823 | ||||
| GenBank mRNA | BC169130.1 | - | 1, 2 | 69 | 6889289 | |
| Gene Symbol: LOC100911746 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003750843.3 | XP_003750891.1 | 1, 2 | 187 | ||
| XM_006246547.2 | XP_006246609.1 | 1, 2 | 817 | |||
| XM_006246548.2 | XP_006246610.1 | 1, 2 | 823 | |||

B9d1
Genomic Map