siRNA Details
| Gene Symbol: Tmem220 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_213341.8 | XP_213341.7 | 3 | 225 | ||
| GenBank mRNA | FQ219853.1 | - | 3 | 163 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem220| Gene Symbol: Tmem220 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_213341.8 | XP_213341.7 | 3 | 225 | ||
| GenBank mRNA | FQ219853.1 | - | 3 | 163 | ||