siRNA Details
Gene Symbol: Tmem220 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_213341.8 | XP_213341.7 | 6 | 552 | ||
GenBank mRNA | FQ219853.1 | - | 6 | 490 | ||
FQ227969.1 | - | 6 | 507 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Tmem220 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_213341.8 | XP_213341.7 | 6 | 552 | ||
GenBank mRNA | FQ219853.1 | - | 6 | 490 | ||
FQ227969.1 | - | 6 | 507 |