siRNA Details
Gene Symbol: Aloxe3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105793.1 | NP_001099263.1 | 13, 14 | 1805 | ||
XM_006246617.2 | XP_006246679.1 | 14, 15 | 2437 | |||
XM_006246619.2 | XP_006246681.1 | 13, 14 | 1877 |
