siRNA Details
| Gene Symbol: Alox12b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039377.1 | NP_001034466.1 | 5 | 722 | ||
| GenBank mRNA | AY903231.1 | - | 5 | 722 | ||
| AY903232.1 | - | 5 | 722 | |||
| AY903233.1 | - | 5 | 722 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Alox12b| Gene Symbol: Alox12b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039377.1 | NP_001034466.1 | 5 | 722 | ||
| GenBank mRNA | AY903231.1 | - | 5 | 722 | ||
| AY903232.1 | - | 5 | 722 | |||
| AY903233.1 | - | 5 | 722 | |||