siRNA Details
Gene Symbol: Eif4a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199372.2 | NP_955404.1 | 7 | 749 | ||
XM_006246624.2 | XP_006246686.1 | 7 | 759 | |||
GenBank mRNA | BC063812.1 | - | 7 | 753 | 6921521 | |
FQ212737.1 | - | 7 | 749 | |||
FQ228869.1 | - | 7 | 751 | |||
FQ233130.1 | - | 7 | 752 |
