siRNA Details
| Gene Symbol: Ctdnep1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100494.1 | NP_001093964.1 | 1, 2 | 313 | ||
| XM_008767804.1 | XP_008766026.1 | 1, 2 | 342 | |||
| GenBank mRNA | BU671232.1 | - | 1, 2 | 313 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ctdnep1| Gene Symbol: Ctdnep1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100494.1 | NP_001093964.1 | 1, 2 | 313 | ||
| XM_008767804.1 | XP_008766026.1 | 1, 2 | 342 | |||
| GenBank mRNA | BU671232.1 | - | 1, 2 | 313 | ||