siRNA Details
| Gene Symbol: Olr1475 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000027.1 | NP_001000027.1 | 1 | 906 | ||
| Gene Symbol: LOC100909438 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003752201.2 | XP_003752249.1 | 1 | 910 | ||

Olr1475
Genomic Map