siRNA Details
| Gene Symbol: Smyd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105810.1 | NP_001099280.1 | 5 | 971 | ||
| XM_006246674.2 | XP_006246736.1 | 5 | 875 | |||
| XM_008767962.1 | XP_008766184.1 | 4 | 1100 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Smyd4| Gene Symbol: Smyd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105810.1 | NP_001099280.1 | 5 | 971 | ||
| XM_006246674.2 | XP_006246736.1 | 5 | 875 | |||
| XM_008767962.1 | XP_008766184.1 | 4 | 1100 | |||