siRNA Details
| Gene Symbol: Prpf8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191590.1 | NP_001178519.1 | 42 | 6917 | ||
| XM_006246892.2 | XP_006246954.1 | 41 | 7118 | |||
| GenBank mRNA | BC099197.1 | - | 7 | 1062 | ||
| FQ211629.1 | - | 4 | 493 | |||
| FQ232414.1 | - | 6 | 1086 | |||

Prpf8
Genomic Map