siRNA Details
Gene Symbol: Ift20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105815.1 | NP_001099285.1 | 5 | 652 | ||
XM_006246935.2 | XP_006246997.1 | 4 | 466 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ift20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105815.1 | NP_001099285.1 | 5 | 652 | ||
XM_006246935.2 | XP_006246997.1 | 4 | 466 |