siRNA Details
| Gene Symbol: Ccl7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007612.1 | NP_001007613.1 | 1, 2 | 165 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccl7| Gene Symbol: Ccl7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007612.1 | NP_001007613.1 | 1, 2 | 165 | ||