siRNA Details
Gene Symbol: Tmem132e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134410.1 | NP_001127882.1 | 2 | 402 | ||
XM_008767965.1 | XP_008766187.1 | 2 | 1814 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Tmem132e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134410.1 | NP_001127882.1 | 2 | 402 | ||
XM_008767965.1 | XP_008766187.1 | 2 | 1814 |